missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Imager White-Light Conversion Screen

Product Code. 13375384
Change view
Click to view available options
For Use With (Equipment):
E-Gel Imager
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
13375384 E-Gel Imager Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13375384 Supplier Invitrogen™ Supplier No. 4473061

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

1 E-Gel Imager White-Light Conversion Screen

Allows visualization of protein gels on the E-Gel Imager

Converts blue light emitted by the E-Gel Imager Blue-Light Base, as well as the UV light emitted by the E-Gel Imager UV-Light Base, to white light to enable use of the powerful and intuitive E-Gel Imager Software package for analysis and quantification of protein gels. The E-Gel Imager White-Light Conversion Screen is compatible with protein gel stains such as SimplyBlue™ Safe, SilverQuest™ Silver, and Coomassie blue stains.

  • A must-have upgrade to E-Gel Imager
  • Ideal for analysis of protein and silver-stained gels
  • Affordable alternative to expensive protein-gel visualization instruments
  • Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 E-Gel™ Imager White-Light Conversion Screen
Store at room temperature
For Use With (Equipment) E-Gel Imager
Product Line E-Gel
Type Imager White-Light Conversion Screen
Shipping Condition Room Temperature

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.