missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Imager System with E-Gel™ Adaptor

Artikelnummer. 11672129
Click to view available options
For Use With (Equipment):
E-Gel Imager
Packungsgröße:
Each
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11672129

Marke: Invitrogen™ 4466613

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Includes

1 x E-Gel™ Imager Camera Hood. 1 x E-Gel™ Imager Adaptor Base. 1 x E-Gel™ Imager software package. 1 x Gel Quant Express software enabling USB dongle. 1 x E-Gel iBase light diffuser

Benötigt

Minimum System Requirements for GelCapture and Gel Quant Express: Processor: Intel™ Core™2 Duo 1.8GHz; Memory: 1Gb Ram; OS: Windows™ XP Professional SP3 (English version) or Windows™ 7 Professional (English version), must have administrator permissions for installation, Monitor: 1024 x 786 minimum resolution, USB: USB 2.0 port for software dongle

Personal imaging system for documenting and analyzing agarose gels and E-Gel™ cassettes

Each E-Gel Imager system includes a sleek and compact camera hood and one or more interchangeable bases along with two powerful software programs.

  • Captures sharp, rich images with the integrated scientific grade camera
  • Captures gel images in real time, even during a run
  • Analyzes gel images using the powerful Gel Quant Express software
  • Powerful and Compact Scientific Grade Camera
  • E-Gel Imager Camera Hood is powerhouse at core of E-Gel Imager system featuring a high resolution, scientific grade camera
  • Captures robust gel images with maximum dynamic range and sensitivity
  • E-Gel Adaptor Base has been designed to provide power to either the E-Gel iBase™ power system coupled to E-Gel Safe Imager kit, or the E-Gel Go Base with its integrated blue light transilluminator so E-Gel cassettes can be run right

  • Gel Compatibility: Agarose Gels, E-Gel™, E-Gel™ 96; Label or Dye: Qdot™ 625, SYBR™
  • Two powerful software tools included with the E-Gel Imager system, GelCapture and Gel Quant Express
  • GelCapture allows to adjust sensitivity and exposure settings to capture the perfect image of your gel
  • Perfect image basic manipulations like inversion or pixel coloration can be performed
  • Gel Quant Express software allows quantitation and analysis via 1D gel analysis or using the express mode
  • GelCapture can be downloaded and utilized on any PC, while Gel Quant Express requires a USB dongle for operation
  • Additional USB dongles can be purchased to use Gel Quant Express on more than one PC simultaneously

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage 1 x E-Gel™ Imager Camera Hood. 1 x E-Gel™ Imager Adaptor Base. 1 x E-Gel™ Imager software package. 1 x Gel Quant Express software enabling USB dongle. 1 x E-Gel Imager light diffuser. 1 x E-Gel Imager universal filter.

Store at room temperature.
For Use With (Equipment) E-Gel Imager
Gel Compatibility E-Gel™, E-Gel™ 48, E-Gel™ 96, Agarose Gels
Product Line E-Gel
Type Imager System with Adapter
System Type E-Gel™ Imager

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt