Learn More
Invitrogen™ E-Gel™ Imager Camera Hood Kit
Description
Includes
Camera Hood - 1 unit: Camera Hood power supply cable (7.5 V) - 1 unit, Camera Hood USB 2.0 cable - 1unit, E-Gel™ Imager Light Diffuser - 1 unit, E-Gel™ Imager Universal Filter - 1 unit, GelCapture Imaging Software, GelQuant Express Quantitation Software - 2 disks, GelQuant Express Software Activation Dongle - 1 unit, Product Quick Reference Card - 1 unit
The E-Gel Camera Hood Kit, when combined with a base, forms the E-Gel Imager System. Each E-Gel Camera Hood Kit includes a compact digital camera mounted in a sleek hood, a universal filter, power and USB cables, and two powerful software applications. There are three bases available for purchase that will work with the E-Gel Camera Hood Kit: a UV Light Base, a Blue Light Base, and an E-Gel Adaptor Base.
- Capture sharp, rich images with the integrated scientific grade digital camera
- Capture gel images in real time, even during a run
- Analyze your gel images using the powerful Gel Quant Express software
- Real Time Image Capture for the First Time
- The E-Gel Imager System is developed to allow real time imaging of E-Gel cassettes during an electrophoresis run
- All three E-Gel Imager System configurations allow you to power and run the E-Gel iBase power system inside the camera hood without having to run wires through a door or under the hood
- By using the E-Gel Camera Hood Kit with the Adaptor Base, you can also run E-Gel Go! cassettes using the Go! Base
- Either way, capture real time images of your agarose gel electrophoresis run like you never have before
- Superb Image Capture and Analysis Software
- There are two powerful software tools included with the E-Gel Camera Hood Kit: GelCapture and Gel Quant Express software
- GelCapture software allows to adjust sensitivity and exposure settings to capture the perfect image of gel
- Once you have that perfect image, basic manipulations like inversion or pixel coloration can be performed
- The Gel Quant Express software allows quantitation and analysis via 1D gel analysis or using the express mode
- GelCapture can be downloaded and run on any PC, while Gel Quant Express requires a USB dongle for operation
- Additional USB dongles can be purchased to use Gel Quant Express on more than one PC simultaneously
- Gel Compatibility: Agarose Gels, E-Gel™, E-Gel™ 48, E-Gel™ 96
- Gel Size: E-Gel™ (8 x 10cm), E-Gel™ 48 (13.5 x 10.8cm), E-Gel™ 96 (13.5 x 10.8cm)
- Mode of Separation: Molecular Weight
- Processor: Intel™ Core™2 Duo 1.8 GHz
- Memory: 1 Gb RAM
- OS: Windows™ XP Professional SP3 (English version) or Windows™ 7 Professional (English version)—must have administrator permissions for installation
- Monitor: 1024 x 786 minimum resolution
- USB: USB 2.0 port for software dongle
- These are the minimum system specifications tested for use of the hardware and software
- Three interchangeable bases that may be purchased separately for use with the E-Gel Camera Hood Kit provide excellent flexibility to use multiple gel stains
- The UV Light Base is best for ethidium bromide ; the Blue Light Base can be used for many SYBR™ dyes like SYBR Safe dye
- The E-Gel Adaptor Base was designed to provide power to either the E-Gel iBase™ power system coupled to the E-Gel Safe Imager kit, or the E-Gel Go! Base with integrated blue light transilluminator, allowing E-Gel cassettes to be run inside the E-Gel Imager
- This feature allows real time image capture while samples are running on the gel
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | Camera Hood - 1 unit Camera Hood power supply cable (7.5 V) - 1 unit Camera Hood USB 2.0 cable - 1unit E-Gel™ Imager Light Diffuser - 1 unit E-Gel™ Imager Universal Filter - 1 unit GelCapture Imaging Software, GelQuant Express Quantitation Software - 2 disks GelQuant Express Software Activation Dongle - 1 unit Product Quick Reference Card - 1 unit |
| For Use With (Equipment) | E-Gel Imager |
| Gel Compatibility | E-Gel™, E-Gel™ 48, E-Gel™ 96, Agarose Gels |
| Gel Size | E-Gel™ 48 (13.5 cm x 10.8 cm), E-Gel™ (8 cm x 10 cm), E-Gel™ 96 (13.5 cm x 10.8 cm) |
| Product Line | E-Gel |
| Type | Imager Camera Hood Kit |
| For Use With | Blue Light Base, UV Light Base, E-Gel™ iBase™, E-Gel™ Adaptor Base, E-Gel™ Go! Base |
| Mode of Separation | Molecular Weight |
| Running Dimension | Horizontal |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
