Learn More
Invitrogen™ E-Gel™ Imager Blue Light Base
Description
Includes
1 x E-Gel™ Imager Blue Light Base
The E-Gel Imager Blue Light Base is one of three interchangeable bases that work with the E-Gel Imager Camera Hood to comprise the E-Gel Imager system. E-Gel Imager Blue Light Base has a built-in safety mechanism that requires the E-Gel Imager Camera Hood to be affixed over the top in order to be operational.
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | 1 x E-Gel™ Imager Blue Light Base. Store at room temperature. |
| For Use With (Equipment) | E-Gel Imager |
| Product Line | E-Gel |
| Type | Imager Blue Light Base |
| Label or Dye | SYBR™ Safe |
| Shipping Condition | Room Temperature |
| System Type | E-Gel™ Imager |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
