missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifications
Specifications
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization |
| Classification | Monoclonal |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize |
| Gene Alias | CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI |
| Regulatory Status | RUO |
| Research Discipline | Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?