missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody (39N7D10) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80834
This item is not returnable.
View return policy
Description
LIF Monoclonal specifically detects LIF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| LIF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 3 - 5 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 5 μg/mL, Immunocytochemistry | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 39N7D10 | |
| Unconjugated | |
| PBS | |
| LIF | |
| A recombinant murine Lif protein containing amino acids 24-203 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction