missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine, MedChemExpress

Product Code. 16458388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458388 10 g 10g
16420808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458388 Supplier Medchem Express Supplier No. HYB0185
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. Lidocaine decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative and has potential for the research of ventricular arrhythmia.

Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. Lidocaine decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative and has potential for the research of ventricular arrhythmia.

Specifications

Chemical Name or Material Lidocaine
Physical Form Solid
CAS 137-58-6
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C14H22N2O
For Use With (Application) Cancer-Kinase/protease
Synonym Lignocaine
Solubility Information DMSO : ≥ 100 mg/mL (426.73 mM) ∣H2O : ≥ 5 mg/mL (21.34 mM)
SMILES O=C(NC1=C(C)C=CC=C1C)CN(CC)CC
Molecular Weight (g/mol) 234.34
Formula Weight 234.34
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.