missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine, MedChemExpress
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. Lidocaine decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative and has potential for the research of ventricular arrhythmia.
Specifications
Specifications
| Chemical Name or Material | Lidocaine |
| Physical Form | Solid |
| CAS | 137-58-6 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C14H22N2O |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | Lignocaine |
| Solubility Information | DMSO : ≥ 100 mg/mL (426.73 mM) ∣H2O : ≥ 5 mg/mL (21.34 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction