missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine, 99.9%, MP Biomedicals™

Product Code. 11494552 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494552 100 g 100g
11404562 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494552 Supplier MP Biomedicals Supplier No. 0219011180
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-58-6 | C14H22N2O | 234.343 g/mol

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
Synonym lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
PubChem CID 3676
ChEBI CHEBI:6456
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)NC1=C(C=CC=C1C)C

Specifications

Color White
Quantity 100 g
Physical Form Powder
Assay Percent Range 99.9%
Formula Weight 234.34
Grade Not Available
Chemical Name or Material Lidocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.