Learn More
Greiner Bio-One Lid for 96-well Plate, Polystyrene, High Profile (9 mm), Sterile
Description
- Clear, polystyrene construction
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
- Suitable for non-TC-treated 96-well microplates
- High lid profile; height: 9 mm
- Sterile
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Sterile |
| No. per Case | 100/Cs. |
| No. per Pack | 1/Pk |
| Plate Compatibility | 96-well Plate |
| Cut-off Corners | Yes |