Learn More
Grant Instruments™ Plastic Lid for Grant Stirred Water Baths
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
Brand:  Grant Instruments™ PL5
Code : 57
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11436467
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
  • Flat or curved moulded plastic
  • Integral handle
  • For use with Grant Stirred Water Baths and heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
Specifications
Specifications
Lid for 5L tank
P5 Plastic Tank
Plastic