Learn More
Grant Instruments™ Plastic Lid for Grant Stirred Water Baths
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
- Flat or curved moulded plastic
- Integral handle
- For use with Grant Stirred Water Baths and heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
Specifications
Specifications
| Description | Lid for 12L tank |
| For Use With (Equipment) | P18 Plastic Tank |
| Material | Plastic |