missing translation for 'onlineSavingsMsg'
Learn More

Licogliflozin, MedChemExpress

Product Code. 16446997
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446997 5 mg 5mg
16440607 50 mg 50mg
16435206 100 mg 100mg
16424278 10 mg 10mg
16403026 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16446997 Supplier Medchem Express Supplier No. HY109092
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Licogliflozin is a sodium glucose cotransporter (SGLT1 and SGLT2) inhibitor.

Licogliflozin is a sodium glucose cotransporter (SGLT1 and SGLT2) inhibitor.

Specifications

Chemical Name or Material Licogliflozin
CAS 1291094-73-9
Color White
Purity Grade Notes Research
Molecular Formula C23H28O7
Quantity 5 mg
Synonym LIK066
Solubility Information DMSO : 100 mg/mL (240.12 mM; Need ultrasonic) ∣H2O : 2 mg/mL (4.80 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](C2=CC(CC3=CC=C4C(OCCO4)=C3)=C(CC)C=C2)O[C@H](CO)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 416.46
Formula Weight 416.46
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.