missing translation for 'onlineSavingsMsg'
Learn More

Licochalcone C, MedChemExpress

Product Code. 16471388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471388 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471388 Supplier Medchem Express Supplier No. HYN0374
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Licochalcone C could inhibit α-glucosidase, with IC50s of <100 nM and 92.43 μM for α-glucosidase and protein tyrosine phosphatase 1B (PTP1B), respectively.

Licochalcone C could inhibit α-glucosidase, with IC50s of <100 nM and 92.43 μM for α-glucosidase and protein tyrosine phosphatase 1B (PTP1B), respectively.

Specifications

Chemical Name or Material Licochalcone C
CAS 144506-14-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22O4
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (98.49 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(O)C=C1)/C=C/C2=CC=C(O)C(C/C=C(C)\C)=C2OC
Molecular Weight (g/mol) 338.4
Formula Weight 338.4
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.