missing translation for 'onlineSavingsMsg'
Learn More

Licochalcone A, MedChemExpress

Product Code. 16468958
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16468958

Brand: Medchem Express HYN0372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Licochalcone A (LCA), a flavonoid isolated, presents obvious anti-cancer effects, displays broad-spectrum inhibition against UDP-glucuronosyltransferases (UGTs). Licochalcone A (LCA) exhibits strong inhibitory effects against UGT1A1, 1A3, 1A4, 1A6, 1A7, 1A9, and 2B7 (both IC50 and Ki values lower than 5 μM) .

Licochalcone A (LCA), a flavonoid isolated, presents obvious anti-cancer effects, displays broad-spectrum inhibition against UDP-glucuronosyltransferases (UGTs). Licochalcone A (LCA) exhibits strong inhibitory effects against UGT1A1, 1A3, 1A4, 1A6, 1A7, 1A9, and 2B7 (both IC50 and Ki values lower than 5 μM) .

Specifications

Chemical Name or Material Licochalcone A
CAS 58749-22-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22O4
Quantity 10 mg
Synonym Licochalcone-A
Solubility Information DMSO : ≥ 31 mg/mL (91.61 mM)
SMILES O=C(C1=CC=C(O)C=C1)/C=C/C2=CC(C(C)(C)C=C)=C(O)C=C2OC
Molecular Weight (g/mol) 338.4
Formula Weight 338.4
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.