Learn More
Macherey Nagel HPLC column analytical LiChrospher 60 RP Select B USP phase L7, eluent in column MeCN/H2O 125mm x 4mm I.D. particle size 5µm Macherey-Nagel
Brand:  Macherey Nagel 728037.40
HPLC column analytical LiChrospher 60 RP Select B USP phase L7, eluent in column MeCN/H2O 125mm x 4mm I.D. particle size 5µm Macherey-Nagel
Code : 16
Additional Details : Weight : 0.01000kg
Product Code. 10012841