Learn More
Berthold Technologies Licence For FB12/Sirius Software
Product Code. 31777426 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Luminescence Assays
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
31777426 Luminescence Assays Each
1 options
Product Code. 31777426 Supplier Berthold Technologies Supplier No. 84113
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Software license to support instrument control and measurement protocols on a Windows PC.
  • Basic PC software allows sample measurement only
  • Data calculations must be performed separately (e.g., in Excel)
  • Sirius 2 license for FB12/Sirius PC software (Microsoft Windows) includes:
  • Single Assay protocol
  • Dual Assay protocol
  • Single Kinetics protocol
  • Multiple Kinetics protocols
  • Cut-off protocol
  • Calculation of replicates
  • Blank subtraction
  • Additional data processing functions
Specifications
Type Software
For Use With (Application) Luminescence Assays
For Use With (Equipment) Sirius 2 LB 9526 Tube Luminometer
System Requirements Windows 10 or Newer