missing translation for 'onlineSavingsMsg'
Learn More

LIAT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen C17orf97
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18265021
View Documents
Novus Biologicals
NBP1-70443
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LIAT1 Polyclonal specifically detects LIAT1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C17orf97
Polyclonal
Rabbit
chromosome 17 open reading frame 97, hypothetical protein LOC400566
C17orf97
IgG
47 kDa
Western Blot
Unconjugated
RUO
400566
Synthetic peptides corresponding to LOC400566(hypothetical gene supported by AK128660) The peptide sequence was selected from the N terminal of LOC400566. Peptide sequence VSLPDFAEIENLANRINESLRWDGILADPEAEKERIRIYKLNRRKRYRCL.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.