Learn More
LIAS Mouse anti-Human, Unconjugated, Clone: 3B5G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67298-1-IG-20UL
This item is not returnable.
View return policy
Description
The LIAS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LIAS. This antibody recognizes human, mouse, rat, pig antigen. The LIAS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| LIAS | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 3B5G7 | |
| Unconjugated | |
| 20 μL | |
| 11019 | |
| IgG1 |