missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LHX9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5107156

Product Code. 17146963

  • £424.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

LHX9 encodes a member of the LIM homeobox gene family of developmentally expressed transcription factors. The encoded protein contains a homeodomain and two cysteine-rich zinc-binding LIM domains involved in protein-protein interactions. The protein is highly similar to a mouse protein that causes gonadal agenesis when inactivated, suggesting a role in gonadal development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LHX9
Polyclonal
Unconjugated
LHX9
3110009O07Rik; BB104635; LH2B; Lhx9; LIM homeobox 9; LIM homeobox 9-like; LIM homeobox protein 9; LIM homeodomain type transcription factor Lhx9; LIM/homeobox protein Lhx9; LIM-homeobox protein 9; LIM-homeodomain type transcription factor 9; si:dkey-121a9.2; zgc:112196
Rabbit
Protein A
RUO
56956
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1.0 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q9NQ69
LHX9
Synthetic peptide directed towards the C-terminal region of human LHX9.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.