Learn More
LHX8 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30722-1-AP-20UL
This item is not returnable.
View return policy
Description
The LHX8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LHX8. This antibody recognizes human antigen. The LHX8 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| LHX8 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Lhx7, LHX8, LIM homeobox 8, LIM homeobox protein 8, LIM/homeobox protein Lhx8 | |
| 20 μL | |
| 431707 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |