missing translation for 'onlineSavingsMsg'
Learn More

Lhx4 Antibody (OTI1C7) [DyLight 680], Novus Biologicals™

Product Code. 30687955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687955 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687955 Supplier Novus Biologicals Supplier No. NBP344979FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lhx4 Monoclonal antibody specifically detects Lhx4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lhx4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI1C7
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CPHD4, FLJ22769, Gsh4, LIM homeobox 4, LIM homeobox protein 4, LIM/homeobox protein Lhx4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-260 of human LHX4 (NP_203129) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89884
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.