Learn More
Invitrogen™ LHX3 Recombinant Rabbit Monoclonal Antibody (24GB3650)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557051
 View more versions of this product
Product Code. 30275948
  • £434.00 / 100µL
Description
LHX3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LHX3
Recombinant Monoclonal
0.315 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P50481, Q9UBR4
LHX3
A synthesized peptide derived from human LHX3 (aa 300-350).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3650
Unconjugated
LHX3
bLhx3b; CPHD3; DKFZp762A2013; Homeobox protein LIM-3; homeobox protein P-LIM; lhx3; Lhx3b; LIM homeo box protein 3; LIM homeobox 3; LIM homeobox protein; LIM homeobox protein 3; LIM homeodomain 3; LIM homeodomain 3 protein b isoform; LIM homeodomain box protein 3; LIM homeodomain protein 3b; LIM/homeobox protein Lhx3; LIM/homeodomain protein LHX3; lim3; Lim-3; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx3; M2-LHX3; mLIM3; mLim-3; Plim; P-LIM
Rabbit
Affinity chromatography
RUO
16871, 170671, 8022
-20°C