missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX3 Rabbit anti-Human, Mouse, Rat, Clone: 0F3J8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | LHX3 |
|---|---|
| Clone | 0F3J8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18354044
|
Novus Biologicals
NBP3-15797-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362322
|
Bio-Techne
NBP3-15797-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LHX3 Monoclonal antibody specifically detects LHX3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| LHX3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8022 | |
| IgG | |
| Affinity purified |
| 0F3J8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CPHD3, DKFZp762A2013, LIM homeobox 3, LIM homeobox protein 3, LIM/homeobox protein Lhx3, LIM/homeodomain protein LHX3, LIM3, M2-LHX3 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human LHX3 (Q9UBR4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title