missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20745-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.Specifications
| LHX3 | |
| Polyclonal | |
| Unconjugated | |
| LHX3 | |
| LHX3, LIM homeobox 3, LIM homeobox protein 3, LIM/homeobox protein Lhx3, LIM3, M2 LHX3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16871, 8022 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50481, Q9UBR4 | |
| Lhx3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction