missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX2 Recombinant Rabbit Monoclonal Antibody (23GB6675), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | LHX2 |
|---|---|
| Clone | 23GB6675 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088002
![]() |
Invitrogen™
MA553092 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088029
|
Invitrogen™
MA553093 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LHX2 Recombinant Monoclonal antibody specifically detects LHX2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| LHX2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P36198, P50458, Q9Z0S2 | |
| 16870, 296706, 9355 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6675 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| A synthesized peptide derived from human LHX2 (1-56AA). | |
| Primary | |
| -20°C | |
| Lhx2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title