missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564870
This item is not returnable.
View return policy
Description
Immunogen sequence: ARDLVYHLNCF TCTTCNKMLT TGDHFGMKDS LVYCRLHFEA LLQGEYPAHF NHADVAAAAA AAAAAKSAGL GSAGANPLGL PYYNGVGTVQ KGRPRKRKSP GPGADLAAYN Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
| LHX2 | |
| Polyclonal | |
| Unconjugated | |
| Lhx2 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50458 | |
| Lhx2 | |
| Recombinant Human LHX2. Recombinant protein control fragment (Product #RP-105675). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction