Learn More
Invitrogen™ LHX2 Monoclonal Antibody (6G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515834
Description
MA5-15834 targets LHX2 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15834 immunogen is purified recombinant fragment of human LHX2 expressed in E. Coli. . MA5-15834 detects LHX2 which has a predicted molecular weight of approximately 44kDa.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
| LHX2 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P50458 | |
| Lhx2 | |
| Purified recombinant fragment of human LHX2 expressed in E. Coli. | |
| RUO | |
| 9355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 6G2 | |
| Unconjugated | |
| Lhx2 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.