missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX2 Antibody (CL6137), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £451.00
Specifications
| Antigen | LHX2 |
|---|---|
| Clone | CL6137 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245701
|
Novus Biologicals
NBP2-61435 |
100 μL |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648768
|
Novus Biologicals
NBP2-61435-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LHX2 Monoclonal specifically detects LHX2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LHX2 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Unconjugated | |
| Mouse | |
| hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2 | |
| LHX2 | |
| IgG1 | |
| Protein A purified |
| CL6137 | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| 9355 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: KRKSPGPGADLAAYNAALS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title