Learn More
LHX2 Antibody (CL6137) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61435UV
Product Code. 30300101
  • £558.00 / 0.10mL
Description
LHX2 Monoclonal antibody specifically detects LHX2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LHX2
Monoclonal
DyLight 350
hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2
This antibody was developed using a synthetic peptide derived from P50458, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience, Stem Cells
9355
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL6137
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified