missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576197
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.
Specifications
| LHR | |
| Polyclonal | |
| Unconjugated | |
| LHCGR | |
| Gpcr19-rs1; HCG receptor; HHG; hypergonadotropic hypogonadism; LCGR; LGR2; LH receptor; LH/CGR; LH/CG-R; LHCGR; LHR; LH-R; LHRHR; LSHR; LSH-R; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; Lutropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; ULG5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16867, 25477, 3973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16235, P22888, P30730 | |
| LHCGR | |
| Recombinant fusion protein of human LHCGR. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction