missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHR Antibody (LHCGR/1417), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53276-20UG
This item is not returnable.
View return policy
Description
LHR Monoclonal specifically detects LHR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LHR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
| Mouse | |
| 82 kDa | |
| 20 μg | |
| GPCR | |
| 3973 | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| LHCGR/1417 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| LHCGR | |
| Recombinant N-terminal fragment of human LHCGR (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction