missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHR Antibody (LHCGR/1415), Novus Biologicals™
Mouse Monoclonal Antibody
£226.00 - £409.00
Specifications
| Antigen | LHR |
|---|---|
| Clone | LHCGR/1415 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286880
|
Novus Biologicals
NBP2-53258-20UG |
20 μg |
£226.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197379
|
Novus Biologicals
NBP2-53258-100UG |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LHR Monoclonal specifically detects LHR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| LHR | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| GPCR | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 3973 | |
| Purified recombinant N-terminal fragment of human LHCGR (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| LHCGR/1415 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
| LHCGR | |
| IgG1 κ | |
| Protein A or G purified | |
| 82 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title