missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15759-1-AP-20UL
This item is not returnable.
View return policy
Description
LHPP is a protein coding gene.Specifications
| LHPP | |
| Polyclonal | |
| Unconjugated | |
| LHPP | |
| hLHPP, LHPP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361663, 64077, 76429 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5I0D5, Q9D7I5, Q9H008 | |
| Lhpp | |
| LHPP Fusion Protein Ag8375 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction