missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHFPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588826
This item is not returnable.
View return policy
Description
Immunogen sequence: SADWLIGKAR SRGGVEPAGP GGGSPEPYHP TLGIYARCIR NPGVQHFQRD TLCGPYAESF GEIASGFWQA T; Positive Samples: Mouse eye; Cellular Location: Membrane, Multi-pass membrane protein.
LHPL2 is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes.
Specifications
| LHFPL2 | |
| Polyclonal | |
| Unconjugated | |
| LHFPL2 | |
| 6030465B15; AI447312; AW050335; I79_004617; KIAA0206; LHFPL tetraspan subfamily member 2 protein; Lhfpl2; LHFP-like protein 2; lipoma HMGIC fusion partner-like 2; lipoma HMGIC fusion partner-like 2 protein; mKIAA0206; vgim | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 218454 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.81 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8BGA2 | |
| LHFPL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-100 of human LHFPL2 (NP_0057701). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction