Learn More
Gibco™ LHC Basal Medium (1X)

Description
LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.
LHC Basal Medium (1X), liquid
Applications:
Formulated for the growth of bronchial epithelial cells with further supplements.
Recommended storage conditions:
2°C to 8°C.
Shelf life:
12 months
Intended use(s)
Research use only
LHC-8 Medium and LHC-9 Medium
Applications:
Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.
Performance and quality testing:
Performance tested in a growth assay using BEAS-2B cells.
Recommended storage conditions:
-5°C to -20°C.
Intended use(s)
Research use only
Reference(s):
Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.
Specifications
Specifications
| Cell Line | Bronchial epithelial cells, IB3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells) |
| Cell Type | Epithelial Cell (Bronchial) |
| Classification | Serum-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | LHC Basal Medium |
| Sterility | Sterile-filtered |
| With Additives | Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate |
| Quantity | 1,000 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.