missing translation for 'onlineSavingsMsg'
Learn More

LH846, MedChemExpress

Product Code. 16459307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459307 5 mg 5mg
16446758 10 mg 10mg
16442837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16459307 Supplier Medchem Express Supplier No. HY15704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LH846 is a selective inhibitor of CKIδ, with an IC50 of 290 nM, and less potently inhibits CKIα and CKIε, with IC50s of 2.5 μM and 1.3 μM, respectively.

LH846 is a selective inhibitor of CKIδ, with an IC50 of 290 nM, and less potently inhibits CKIα and CKIε, with IC50s of 2.5 μM and 1.3 μM, respectively.

Specifications

Chemical Name or Material LH846
Physical Form Powder
CAS 639052-78-1
Color Pale Purple
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H13ClN2OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 45 mg/mL (142.04 mM)
SMILES O=C(NC1=NC2=CC(Cl)=C(C)C=C2S1)CC3=CC=CC=C3
Molecular Weight (g/mol) 316.81
Formula Weight 316.81
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.