missing translation for 'onlineSavingsMsg'
Learn More

LH846, MedChemExpress

Product Code. 16446758
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446758

Brand: Medchem Express HY15704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LH846 is a selective inhibitor of CKIδ, with an IC50 of 290 nM, and less potently inhibits CKIα and CKIε, with IC50s of 2.5 μM and 1.3 μM, respectively.

LH846 is a selective inhibitor of CKIδ, with an IC50 of 290 nM, and less potently inhibits CKIα and CKIε, with IC50s of 2.5 μM and 1.3 μM, respectively.

Specifications

Chemical Name or Material LH846
Physical Form Powder
CAS 639052-78-1
Color Pale Purple
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H13ClN2OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 45 mg/mL (142.04 mM)
SMILES O=C(NC1=NC2=CC(Cl)=C(C)C=C2S1)CC3=CC=CC=C3
Molecular Weight (g/mol) 316.81
Formula Weight 316.81
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.