missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LH beta Monoclonal antibody specifically detects LH beta in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | LH beta |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rLHb/1613 |
| Conjugate | DyLight 680 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human LH beta protein (Uniprot: P01229) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?