missing translation for 'onlineSavingsMsg'
Learn More

LH beta Antibody [PE], Novus Biologicals™

Product Code. 30731836 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731836 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731836 Supplier Novus Biologicals Supplier No. AF8016P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

LH beta Polyclonal antibody specifically detects LH beta in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LH beta
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta
Host Species Sheep
Immunogen E. coli-derived recombinant human LHβ, Ser21-Leu141, Accession # P01229
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3972
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.