missing translation for 'onlineSavingsMsg'
Learn More

LGX 818, TRC

Product Code. 30010374
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010374 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010374 Supplier Toronto Research Chemicals Supplier No. TRCL3976555MG
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1269440-17-6
Molecular Formula C22 H27 Cl F N7 O4 S
Quantity 5 mg
Synonym Carbamic acid, N-[(1S)-2-[[4-[3-[5-chloro-2-fluoro-3-[(methylsulfonyl)amino]phenyl]-1-(1-methylethyl)-1H-pyrazol-4-yl]-2-pyrimidinyl]amino]-1-methylethyl]-, methyl ester, (S)-Methyl [1-[[4-[3-[5-chloro-2-fluoro-3-(methylsulfonamido)phenyl]-1-isopropyl-1H-pyrazol-4-yl]pyrimidin-2-yl]amino]propan-2-yl]carbamate, Encorafenib, LGX 818, NVP-LGX 818NXA, N-[(1S)-2-[[4-[3-[5-Chloro-2-fluoro-3-[(methylsulfonyl)amino]phenyl]-1-(1-methylethyl)-1H-pyrazol-4-yl]-2-pyrimidinyl]amino]-1-methylethyl]-carbamic Acid Methyl Ester, LGX 818, N-[(1S)-2-[[4-[3-[5-Chloro-2-fluoro-3-[(methylsulfonyl)amino]phenyl]-1-(1-methylethyl)-1H-pyrazol-4-yl]-2-pyrimidinyl]amino]-1-methylethyl]-carbamic Acid Methyl Ester, (S)-Methyl [1-[[4-[3-[5-chloro-2-fluoro-3-(methylsulfonamido)phenyl]-1-isopropyl-1H-pyrazol-4-yl]pyrimidin-2-yl]amino]propan-2-yl]carbamate
InChI Formula InChI=1S/C22H27ClFN7O4S/c1-12(2)31-11-16(17-6-7-25-21(28-17)26-10-13(3)27-22(32)35-4)20(29-31)15-8-14(23)9-18(19(15)24)30-36(5,33)34/h6-9,11-13,30H,10H2,1-5H3,(H,27,32)(H,25,26,28)/t13-/m0/s1
SMILES COC(=O)N[C@@H](C)CNc1nccc(n1)c2cn(nc2c3cc(Cl)cc(NS(=O)(=O)C)c3F)C(C)C
IUPAC Name methyl N-[(1S)-2-[[4-[3-[5-chloro-2-fluoro-3-(methanesulfonamido)phenyl]-1-isopropyl-pyrazol-4-yl]pyrimidin-2-yl]amino]-1-methyl-ethyl]carbamate
Molecular Weight (g/mol) 540.01
Formula Weight 539.1518
Chemical Name or Material Encorafenib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.