missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGTN Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50038PCP
This item is not returnable.
View return policy
Description
LGTN Polyclonal antibody specifically detects LGTN in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| LGTN | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| eukaryotic translation initiation factor 2D, HCA56eIF2d, Hepatocellular carcinoma-associated antigen 56, LGTNLigatin, ligatin | |
| The immunogen for this product maps to a region between residue 350 and 400 of human eIF2D using the numbering given in entry NP_008824.2 (GeneID 1939). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 1939 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction