missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr5/GPR49 Antibody (CL13579) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24571APC
This item is not returnable.
View return policy
Description
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Lgr5/GPR49 | |
| Monoclonal | |
| APC | |
| FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| This antibody was developed using a synthetic peptide derived from O75473, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway | |
| 8549 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL13579 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction