missing translation for 'onlineSavingsMsg'
Learn More

Lgr5/GPR49 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18341467 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341467 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341467 Supplier Novus Biologicals Supplier No. NLS1235AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lgr5/GPR49 Polyclonal specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5/GPR49
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O75473
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Gene Symbols LGR5
Host Species Rabbit
Immunogen Synthetic peptide [KLH conjugated] made to the 2nd cytoplasmic loop of human GPR49/LGR5. [UniProt# O75473]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Test Specificity Human. Immunogen sequence has 81% identity with mouse.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.