missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109908
This item is not returnable.
View return policy
Description
Immunogen sequence: HLESLTLTGT KISSIPNNLC QEQKMLRTLD LSYNNIRDLP SFNGCHALEE ISLQRNQIYQ IKEGTFQGLI SLRILDLSRN LIHEIHSRAF ATLGPITNLD VSFNELTSFP TEGLNGLNQL KLVGNFKLKE ALAAKDFVNL RSLSVPYAYQ CCAFWGCDSY ANLNTEDNSL QDHSVAQEKG TADAANVTST LENEEHSQII IHCTPSTGAF KPCEYLLGSW M.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR LGR4 is a member of this family (subfamily Orphan-A). Its expression has been documented in human brain, adrenal, heart, kidney, liver, skeletal muscle, ovary, pancreas, placenta, prostate, spleen, stomach, testis, thyroid, and spinal cord.
Specifications
| LGR4 | |
| Polyclonal | |
| Unconjugated | |
| LGR4 | |
| 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107515, 286994, 55366 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| A2ARI4, Q9BXB1, Q9Z2H4 | |
| LGR4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-540 of human LGR4 (NP_060960.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction