missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr4/GPR48 Antibody (852229) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7750JF525
This item is not returnable.
View return policy
Description
Lgr4/GPR48 Monoclonal antibody specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,Immunocytochemistry,CyTOF
Specifications
| Lgr4/GPR48 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
| NS0 mouse myeloma cell line transfected with human Lgr4/GPR48, Met1-Asp951, Accession # Q9BXB1 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| 852229 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 55366 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction