missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848612
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848612 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848612 Supplier Invitrogen™ Supplier No. PA520434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is MDA-MB-361 cell lysate. PA5-20434 can be used with blocking peptide PEP-0551.

Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DHX58
Gene Accession No. Q96C10, Q99J87
Gene Alias B430001I08Rik; D11LGP2; D11lgp2e; DEXH (Asp-Glu-X-His) box polypeptide 58; DEXH-box helicase 58; DHX58; Lgp2; LPG2; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; Probable ATP-dependent RNA helicase DHX58; protein D11Lgp2; Protein D11Lgp2 homolog; RGD1310093; RIG-I-like receptor 3; RIG-I-like receptor Lgp2; RLR; RLR-3; RNA helicase LGP2
Gene Symbols DHX58
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus of human LGP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303538, 79132, 80861
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.