Learn More
Invitrogen™ LGMN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587990
Description
Positive Samples: Mouse lung, Mouse liver; Cellular Location: Lysosome Immunogen sequence: GGKHWVVIVA GSNGWYNYRH QADACHAYQI IHRNGIPDEQ IVVMMYDDIA YSEDNPTPGI VINRPNGTDV YQGVPKDYTG EDVTPQNFLA VLRGDAEAVK GIGSGKVLKS GPQDHVFIYF TDHGSTGILV FPNEDLHVKD LNETIHYMYK HKMYRKMVFY IEACESGSMM NHLPDNINVY ATTAANPRES SYACYYDEKR STYLGDWYSV NWMEDSDVED LTKETLHKQY HLVKSHTNTS HVMQYGNKTI STMKVMQFQG MKR.
This gene encodes a cysteine protease that has a strict specificity for hydrolysis of asparaginyl bonds. This enzyme may be involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation in the lysosomal/endosomal systems. Enzyme activation is triggered by acidic pH and appears to be autocatalytic. Protein expression occurs after monocytes differentiate into dendritic cells. A fully mature, active enzyme is produced following lipopolysaccharide expression in mature dendritic cells. Overexpression of this gene may be associated with the majority of solid tumor types. This gene has a pseudogene on chromosome 13. Several alternatively spliced transcript variants have been described, but the biological validity of only two has been determined.
Specifications
| LGMN | |
| Polyclonal | |
| Unconjugated | |
| LGMN | |
| AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19141, 5641, 63865 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.01 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O89017, Q99538, Q9R0J8 | |
| LGMN | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.