Learn More
LGMN Mouse anti-Human, Unconjugated, Clone: 1F2B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67017-1-IG-150UL
This item is not returnable.
View return policy
Description
The LGMN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LGMN. This antibody recognizes human antigen. The LGMN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| LGMN | |
| Monoclonal | |
| 960 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F2B12 | |
| Unconjugated | |
| 150 μL | |
| 5641 | |
| IgG1 |