missing translation for 'onlineSavingsMsg'
Learn More

Lgi4 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18431525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431525 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431525 Supplier Novus Biologicals Supplier No. NBP176381FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lgi4 Polyclonal antibody specifically detects Lgi4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgi4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias leucine-rich glioma-inactivated gene 4, Leucine-rich glioma-inactivated protein 4, leucine-rich repeat LGI family member 4, leucine-rich repeat LGI family, member 4, LGI1-like protein 3, LGIL3
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human LGI4. The immunogen is located within amino acids 470 - 520 of LGI4. Amino Acid Squence: EPDKGLLEPLQELGPPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 163175
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.